3-Brom-5-nitrobenzoesäure

Lieferant: SIA ENAMINE
Achtung

EN300106819.1G
ENAMEN300106819.1GEA 0 EUR
ENAMEN300106819.1G
3-Brom-5-nitrobenzoesäure
3-Bromo-5-nitrobenzoic acid
Formel: C₇H₄BrNO₄
Molecular Weight: 246,02 g/mol
MDL: MFCD00100098
CAS-Nummer: 6307-83-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR