5-Nitro-2-thiophencarbonitril

Lieferant: SIA ENAMINE

Synonyme: 5-Nitrothiophene-2-carbonitrile

EN300106869.1G
ENAMEN300106869.1GEA 0 EUR
ENAMEN300106869.1G
5-Nitro-2-thiophencarbonitril
5-Nitro-2-thiophencarbonitril
Formel: C₅H₂N₂O₂S
Molecular Weight: 154,15 g/mol
MDL: MFCD00052401
CAS-Nummer: 16689-02-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR