Methyl-3-formylbenzoat

Lieferant: SIA ENAMINE

Synonyme: Methyl 3-formylbenzoate

EN300106923.1G
ENAMEN300106923.1GEA 0 EUR
ENAMEN300106923.1G
Methyl-3-formylbenzoat
Methyl-3-formylbenzoat
Formel: C₉H₈O₃
Molecular Weight: 164,1600 g/mol
MDL: MFCD00189379
CAS-Nummer: 52178-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR