5-Fluoroxindol

Lieferant: SIA ENAMINE

Synonyme: 5-Fluoroindolin-2-one, 5-Fluoro-2-oxoindoline, 5-Fluoro-2-oxindole

EN300106959.1G
ENAMEN300106959.1GEA 0 EUR
ENAMEN300106959.1G
5-Fluoroxindol
5-Fluoroxindol
Formel: C₈H₆FNO
Molecular Weight: 151,14 g/mol
MDL: MFCD02179598
CAS-Nummer: 56341-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR