2-Brom-4-chlorbenzaldehyd

Lieferant: SIA ENAMINE

EN300107015.1G
ENAMEN300107015.1GEA 0 EUR
ENAMEN300107015.1G
2-Brom-4-chlorbenzaldehyd
2-Bromo-4-chlorobenzaldehyde
Formel: C₇H₄BrClO
MDL: MFCD09056792
CAS-Nummer: 84459-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR