3,5-Dimethyl-4-fluorbenzonitril

Lieferant: SIA ENAMINE

EN300107063.1G
ENAMEN300107063.1GEA 0 EUR
ENAMEN300107063.1G
3,5-Dimethyl-4-fluorbenzonitril
3,5-Dimethyl-4-fluorbenzonitril
Formel: C₉H₈FN
Molecular Weight: 149,1700 g/mol
MDL: MFCD09832248
CAS-Nummer: 867367-02-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR