4-Chlor-3-(trifluormethyl)benzoesäure

Lieferant: SIA ENAMINE

Synonyme: 4-Chlor-α,α,α-trifluor-m-toluylsäure

EN300107294.1G
ENAMEN300107294.1GEA 0 EUR
ENAMEN300107294.1G
4-Chlor-3-(trifluormethyl)benzoesäure
4-Chloro-3-(trifluoromethyl)benzoic acid
Formel: C₈H₄ClF₃O₂
Molecular Weight: 224,57 g/mol
MDL: MFCD01631468
CAS-Nummer: 1737-36-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR