4-Brom-2-nitrobenzaldehyd

Lieferant: SIA ENAMINE

EN300107308.1G
ENAMEN300107308.1GEA 0 EUR
ENAMEN300107308.1G
4-Brom-2-nitrobenzaldehyd
4-Bromo-2-nitrobenzaldehyde
Formel: C₇H₄BrNO₃
MDL: MFCD00463687
CAS-Nummer: 5551-12-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR