5-Brom-4,6-dichlorpyrimidin

Lieferant: SIA ENAMINE

EN300107356.1G
ENAMEN300107356.1GEA 0 EUR
ENAMEN300107356.1G
5-Brom-4,6-dichlorpyrimidin
5-Bromo-4,6-dichloropyrimidine
Formel: C₄HBrCl₂N₂
MDL: MFCD00047379
CAS-Nummer: 68797-61-5
EINECS: 272-305-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR