7-(Trifluormethyl)isatin
Lieferant: SIA ENAMINE
EN30010741.1G
ENAMEN30010741.1GEA
0
EUR
ENAMEN30010741.1G
7-(Trifluormethyl)isatin
7-(Trifluormethyl)isatin
Formel:
C₉H₄F₃NO₂ Molecular Weight: 215,1300 g/mol |
MDL:
MFCD01248408 CAS-Nummer: 391-12-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...