2-Chlor-6-methyl-5-nitropyridin

Lieferant: SIA ENAMINE

Synonyme: 6-Chlor-2-methyl-3-nitropyridin, 2-Chlor-5-nitro-6-picolin, 6-Chlor-3-nitro-2-picolin

EN300107675.1G
ENAMEN300107675.1GEA 0 EUR
ENAMEN300107675.1G
2-Chlor-6-methyl-5-nitropyridin
2-Chloro-6-methyl-5-nitropyridine
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172,57 g/mol
MDL: MFCD00234160
CAS-Nummer: 22280-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR