5-Iod-o-toluylsäure
Lieferant: SIA ENAMINE
Synonyme:
5-lodo-2-methylbenzoic acid, 5-Iodo-2-methylbenzoic acid
EN300107760.1G
ENAMEN300107760.1GEA
0
EUR
ENAMEN300107760.1G
5-Iod-o-toluylsäure
5-Iod-o-toluylsäure
Formel:
C₈H₇IO₂ |
MDL:
MFCD01570258 CAS-Nummer: 54811-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...