2-Chlor-5-iodbenzoesäure

Lieferant: SIA ENAMINE

EN300107761.1G
ENAMEN300107761.1GEA 0 EUR
ENAMEN300107761.1G
2-Chlor-5-iodbenzoesäure
2-Chlor-5-iodbenzoesäure
Formel: C₇H₄ClIO₂
Molecular Weight: 282,46 g/mol
MDL: MFCD00079731
CAS-Nummer: 19094-56-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR