2-Fluor-5-iodbenzoesäure

Lieferant: SIA ENAMINE
Achtung

EN300107762.1G
ENAMEN300107762.1GEA 0 EUR
ENAMEN300107762.1G
2-Fluor-5-iodbenzoesäure
2-Fluor-5-iodbenzoesäure
Formel: C₇H₄FIO₂
Molecular Weight: 266,01 g/mol
MDL: MFCD03094517
CAS-Nummer: 124700-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR