4-Fluor-3-iodbenzoesäure

Lieferant: SIA ENAMINE

EN300107765.1G
ENAMEN300107765.1GEA 0 EUR
ENAMEN300107765.1G
4-Fluor-3-iodbenzoesäure
4-Fluoro-3-iodobenzoic acid
Formel: C₇H₄FIO₂
MDL: MFCD03788556
CAS-Nummer: 403-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR