3-Iod-4-methoxybenzoesäure

Lieferant: SIA ENAMINE

Synonyme: 3-Iodo-p-anisic acid

EN300107772.1G
ENAMEN300107772.1GEA 0 EUR
ENAMEN300107772.1G
3-Iod-4-methoxybenzoesäure
3-Iod-4-methoxybenzoesäure
Formel: C₈H₇IO₃
Molecular Weight: 278,05 g/mol
MDL: MFCD00017347
CAS-Nummer: 68507-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR