3-Ethoxy-4-hydroxybenzonitril

Lieferant: SIA ENAMINE

EN30010778.1G
ENAMEN30010778.1GEA 0 EUR
ENAMEN30010778.1G
3-Ethoxy-4-hydroxybenzonitril
3-Ethoxy-4-hydroxybenzonitril
Formel: C₉H₉NO₂
MDL: MFCD01123034
CAS-Nummer: 60758-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR