5'-Chlor-2'-hydroxypropiophenon

Lieferant: SIA ENAMINE

EN30010779.1G
ENAMEN30010779.1GEA 0 EUR
ENAMEN30010779.1G
5'-Chlor-2'-hydroxypropiophenon
5'-Chlor-2'-hydroxypropiophenon
Formel: C₉H₉ClO₂
MDL: MFCD00218579
CAS-Nummer: 2892-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR