(6-Morpholino-3-pyridinyl)methanol

Lieferant: SIA ENAMINE

EN300107838.1G
ENAMEN300107838.1GEA 0 EUR
ENAMEN300107838.1G
(6-Morpholino-3-pyridinyl)methanol
(6-Morpholino-3-pyridinyl)methanol
Formel: C₁₀H₁₄N₂O₂
MDL: MFCD03086115
CAS-Nummer: 388088-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR