6-Chlorimidazo[1,2-b]pyridazin

Lieferant: SIA ENAMINE

EN300107839.1G
ENAMEN300107839.1GEA 0 EUR
ENAMEN300107839.1G
6-Chlorimidazo[1,2-b]pyridazin
6-Chlorimidazo[1,2-b]pyridazin
Formel: C₆H₄ClN₃
MDL: MFCD07778345
CAS-Nummer: 6775-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR