5-Iod-1,3-benzodioxol

Lieferant: SIA ENAMINE

Synonyme: 5-Iodobenzo[d][1,3]dioxole

EN300107850.1G
ENAMEN300107850.1GEA 0 EUR
ENAMEN300107850.1G
5-Iod-1,3-benzodioxol
5-Iod-1,3-benzodioxol
Formel: C₇H₅IO₂
Molecular Weight: 248,0179 g/mol
MDL: MFCD00079766
CAS-Nummer: 5876-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR