(2-Fluor-4-nitrophenyl)essigsäure

Lieferant: SIA ENAMINE

Synonyme: 2-(2-Fluoro-4-nitrophenyl)acetic acid

EN300107917.1G
ENAMEN300107917.1GEA 0 EUR
ENAMEN300107917.1G
(2-Fluor-4-nitrophenyl)essigsäure
(2-Fluor-4-nitrophenyl)essigsäure
Formel: C₈H₆FNO₄
Molecular Weight: 199,1360 g/mol
MDL: MFCD11041422
CAS-Nummer: 315228-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR