4-Cyan-2-fluorbenzaldehyd

Lieferant: SIA ENAMINE

Synonyme: 3-Fluoro-4-formylbenzonitrile

EN300107978.1G
ENAMEN300107978.1GEA 0 EUR
ENAMEN300107978.1G
4-Cyan-2-fluorbenzaldehyd
4-Cyano-2-fluorobenzaldehyde
Formel: C₈H₄FNO
Molecular Weight: 149,1200 g/mol
MDL: MFCD07368786
CAS-Nummer: 105942-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR