2-Brom-5-sulfamoylbenzoesäure

Lieferant: SIA ENAMINE

EN30010799.1G
ENAMEN30010799.1GEA 0 EUR
ENAMEN30010799.1G
2-Brom-5-sulfamoylbenzoesäure
2-Brom-5-sulfamoylbenzoesäure
Formel: C₇H₆BrNO₄S
MDL: MFCD06355953
CAS-Nummer: 22361-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR