Methyl-4-amino-2-fluorbenzoat

Lieferant: SIA ENAMINE

EN300108007.1G
ENAMEN300108007.1GEA 0 EUR
ENAMEN300108007.1G
Methyl-4-amino-2-fluorbenzoat
Methyl-4-amino-2-fluorbenzoat
Formel: C₈H₈FNO₂
MDL: MFCD09037917
CAS-Nummer: 73792-08-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR