2,5-Dibrom-3-nitropyridin

Lieferant: SIA ENAMINE

EN300108015.1G
ENAMEN300108015.1GEA 0 EUR
ENAMEN300108015.1G
2,5-Dibrom-3-nitropyridin
2,5-Dibromo-3-nitropyridine
Formel: C₅H₂Br₂N₂O₂
MDL: MFCD09266223
CAS-Nummer: 15862-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR