4-Chlor-2-benzo[b]thiophencarbonsäure

Lieferant: SIA ENAMINE

EN30010812.1G
ENAMEN30010812.1GEA 0 EUR
ENAMEN30010812.1G
4-Chlor-2-benzo[b]thiophencarbonsäure
4-Chlor-2-benzo[b]thiophencarbonsäure
Formel: C₉H₅ClO₂S
Molecular Weight: 212,65 g/mol
MDL: MFCD06655155
CAS-Nummer: 23967-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR