1,1,1-Trifluor-3-phenylaceton
Lieferant: SIA ENAMINE
Synonyme:
3-Phenyl-1,1,1-trifluoropropan-2-one, 1,1,1-Trifluoro-3-phenyl-2-propanone, (3,3,3-Trifluor-2-propanoyl)benzene, 1,1,1-Trifluoro-3-phenylpropan-2-one
EN300108150.1G
ENAMEN300108150.1GEA
0
EUR
ENAMEN300108150.1G
1,1,1-Trifluor-3-phenylaceton
1,1,1-Trifluor-3-phenylaceton
Formel:
C₉H₇F₃O Molecular Weight: 188,15 g/mol |
MDL:
MFCD00068171 CAS-Nummer: 350-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...