2-Brom-3-nitro-4-picolin

Lieferant: SIA ENAMINE

Synonyme: 2-Brom-4-methyl-3-nitropyridin

EN300108151.1G
ENAMEN300108151.1GEA 0 EUR
ENAMEN300108151.1G
2-Brom-3-nitro-4-picolin
2-Brom-3-nitro-4-picolin
Formel: C₆H₅BrN₂O₂
Molecular Weight: 217,02 g/mol
MDL: MFCD04112527
CAS-Nummer: 23056-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR