Conductivity Calibration Solution
Lieferant: Endress+Hauser
CLY11-A
ENDRCLY11-AEA 342 EUR
ENDRCLY11-A
Conductivity Calibration Solution
Standards Leitfähigkeits-Standards
Precision solution referred to SRM of NIST.
With temperature table for qualified calibration of conductivity measuring systems acc. to ISO.