Digital pH Sensor, Memosens CPS71E
Lieferant: Endress+Hauser
CPS71E-1009/0 CPS71E-1265 CPS71E-BA7BTB2
ENDRCPS71E-1009/0EA 497 EUR
ENDRCPS71E-1009/0 ENDRCPS71E-1265 ENDRCPS71E-BA7BTB2
Digital pH Sensor, Memosens CPS71E
Elektroden pH-Elektroden
With its contamination-resistant reference system, Memosens CPS71E is a digital pH sensor for highly contaminated or toxic media such as strong acids or bases.
  • The special ion trap of the electrode prevents poisoning of the junction and the reference, offers resistance to strong acids and bases and ensures a longer service life of the sensor
  • The optional pressurized reference ensures reliable measurements in blocking media
  • Optional upside-down mounting for flexible installation
  • Inductive cable connection and non-contact signal transmission improve process
More Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable