Handles for device carriers
Lieferant: ENTEGRIS
A07-0215 A01-0215 PA031-0603 A031-0215
ENTEA07-0215EA 0 EUR
ENTEA07-0215 ENTEA01-0215 ENTEPA031-0603 ENTEA031-0215
Handles for device carriers
Wafer-Handling-Zubehör Wafer-Carrier
Squeeze style handles.
  • Allows easy engagement and disengagement from the carrier
  • Attaches to the notches in both end walls of the carrier
  • Applying pressure to the center of the two cross members and squeezing causes the struts to separate
  • For use with various miscellaneous carriers
  • Lead time: eight weeks
Stick handles
Rectangular or square styles
For use with LCD carriers, miscellaneous carriers, wafer baskets and labware trays
Lead time: eight weeks
Bestellen
TECHNISCHE DATEN
Environmentally Preferable