Mask packages, B 80 series
Lieferant: ENTEGRIS
B8030-0211 B8050-1611 B7950-0100 B8050-0600 B8060-0611 B8060-0700 B8060-2011 B8060-2400 B8070-0300
ENTEB8030-0211EA 0 EUR
ENTEB8030-0211 ENTEB8050-1611 ENTEB7950-0100 ENTEB8050-0600 ENTEB8060-0611 ENTEB8060-0700 ENTEB8060-2011 ENTEB8060-2400 ENTEB8070-0300
Mask packages, B 80 series
Wafer-Handling-Zubehör Wafer-Carrier
Provide excellent security for masks and reticles with pellicles.
  • High impact resistance
  • Stackable
  • Easy to open and close
  • Seal securely to provide a protective environment inside