Dispensing head, Sentry®
Lieferant: ENTEGRIS
DSU-00-01-00
ENTEDSU-00-01-00EA 0 EUR
ENTEDSU-00-01-00
Dispensing head, Sentry®
Pumpen Fass/Transferpumpen
Designed for use with FluoroPure® single-use HDPE blowmolded drums, this single-use dispense head provides dispensing functions in a single connection.
  • Reliable, high purity o-ring-free dispense connection without chemical key coding
  • All PFA wetted materials provide corrosion resistance and high purity
  • Fill and dispense port comes standard with a 1" Flaretek® flared connection