Manifold, Impact® ST
Lieferant: ENTEGRIS
DZUM044C1
ENTEDZUM044C1EA 0 EUR
ENTEDZUM044C1
Manifold, Impact® ST
Verteiler
Stainless steel manifold for use with Impact® Plus disposable filters.
  • Rapid filter changeout with minimal dripping: no tools and no draining required
  • Compact design provides easy installation or retrofit into existing tools