GateKeeper® GPU 20M 12, 16R Gas Purifiers
Lieferant: ENTEGRIS
GPUS20MFOP12R0
ENTEGPUS20MFOP12R0EA 0 EUR
ENTEGPUS20MFOP12R0
GateKeeper® GPU 20M 12, 16R Gas Purifiers
Filter Gasfilter
GateKeeper® GPU gas purifiers are designed to improve purity consistency, lifetime, and final purity in microelectronics applications where high-purity gas is critical.
  • Purity control below part-per-billion (ppb) levels
  • Low pressure drop through the purifier
  • Regenerable - eliminates disposal issues and costs
  • Long lifetime - improves cost of ownership
  • Insensitive to orientation (horizontal / vertical)
Uses ambient temperature purification - no heat or power required to operate.
Applications include semiconductor, data storage, flat panel display, LED and solar.
Standards: Manufactured in an ISO® 9001:2015 facility.