GateKeeper® GPU CO Media Gas Purifier

Lieferant: ENTEGRIS
GPUS3000FCR08R
ENTEGPUS3000FCR08REA 0 EUR
ENTEGPUS3000FCR08R
GateKeeper® GPU CO Media Gas Purifier
Filter Gasfilter
Entegris GateKeeper® GPU gas purifiers were designed to improve purity consistency, lifetime, and final purity in microelectronics applications where high-purity gas is critical. Leveraging our expertise in material science, we have developed a new generation of CO gas purification products that deliver unsurpassed value and performance.

  • Purity control below part-per-billion (ppb) levels
  • Low pressure drop through the purifier
  • Long lifetime - improves cost of ownership
  • Uses ambient temperature purification - no heat or power required to operate
  • Insensitive to orientation (horizontal/vertical)

Applications: Semiconductor, data storage, flat panel display, LED, solar.

Standards: Manufactured in an ISO® 9001:2015 facility.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR