Multiple wafer shipping boxes for 2,5" and smaller wafer handling

Lieferant: ENTEGRIS
H6620-0009
ENTEH6620-0009EA 0 EUR
ENTEH6620-0009
Multiple wafer shipping boxes for 2,5" and smaller wafer handling
Wafer-Handling-Zubehör Wafer-Versandbehälter
  • Full-depth wafer protection
  • Positive cover alignment
  • Individual wafer support springs
  • Stackable
  • Assembly overall size: 160×60×67 mm
  • Pitch: 4,76 mm
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR