Mask packages, H 92 series
Lieferant: ENTEGRIS
H9260251061116
ENTEH9260251061116EA 0 EUR
ENTEH9260251061116
Mask packages, H 92 series
Wafer-Handling-Zubehör Wafer-Carrier
Provides excellent security for masks or reticles with pellicles.
  • High impact resistance
  • Stackable
  • Easy to open and close
  • Seal securely to provide a protective environment inside
  • Label wall for identification label