Jig trays
Lieferant: ENTEGRIS
P44-JIG-101-4
ENTEP44-JIG-101-4EA 0 EUR
ENTEP44-JIG-101-4
Jig trays
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Patented JEDEC thick profile jig tray.
  • Holds three H44 or P44 trays
  • Maximum temperature: 180 °C
  • Order in multiples of five trays