Planarcore® PVA Brush
Lieferant: ENTEGRIS
PVP0L0TR4
ENTEPVP0L0TR4EA 0 EUR
ENTEPVP0L0TR4
Planarcore® PVA Brush
Bürsten
Planarcore® PVA Brush is designed to deliver superior performance and wafer-to-wafer uniformity in post CMP wafer cleaning applications.
  • High-Purity PVA
  • Molded through the core construction
  • Eliminates alignment and gapping problems, increasing system throughput
  • Close molded technology
  • Reduces downtime on tool
  • Decreases defectivity on wafer
The unique molded-through-the-core technology provides absolute adhesion of the PVA to the brush core, unlike standard PVA products that are merely friction fitted to the core.