Membrane disc filters, Savana®
Lieferant: ENTEGRIS
SVLD047D100A
ENTESVLD047D100AEA 0 EUR
ENTESVLD047D100A
Membrane disc filters, Savana®
Filter Membranfilter
These filter disc series with non-woven PP media can be used for simple, quick and efficient filtration of fluids used in laboratory, pilot and small scale applications. Can be used for clarification of acids, bases and solvents.
  • Can be used in cartridge filters to allow for consistent scale up of products
  • Pressure rating depends on holder rating