Fluoroline® Ultrapure PFA tubings
Lieferant: ENTEGRIS
UAT1000-062
ENTEUAT1000-062EA 0 EUR
ENTEUAT1000-062
Fluoroline® Ultrapure PFA tubings
Schläuche Transferschlauch
Made of PFA, DuPont 450.
  • Compliant to SEMI Standard F57-0301, the specification for polymer components used in ultrapure water and liquid chemical distribution systems
  • Made of 100% virgin high purity material to provide the lowest extractable ionic and organic levels in PFA materials
  • Tight and consistent tolerances, along with lot-to-lot consistency, are maintained using continuous on-line measurements of the FluoroLine® tubing Ext. Ø. and wall thickness
  • Tubing is produced with specially designed extruders and extrusion processes
Permanent serialised laser marking is a nondestructive and noncontaminating method to identify tube material and Ext. Ø. It also allows for 100% traceability to a specific material lot, production date and production inspection record.