[pThr183/Tyr185]Jnk1/2 ELISA kit
Lieferant: ENZO LIFE SCIENCES
ADI900106
ENZOADI900106EA 750 EUR
ENZOADI900106
[pThr183/Tyr185]Jnk1/2 ELISA kit
Tests ELISAs
For the quantitative determination of [pThr183/Tyr185]Jnk1/2 in cell lysates of human, mouse, and rat origin.
The [pThr183/Tyr185]Jnk1/2 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours. Absorbance is read at 450nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Bestellinformation: Contains Microtiter plate, Conjugate, Antibody, Assay buffer 4 concentrate, Wash buffer concentrate, Standard, TMB Substrate, RIPA Cell lysis buffer 2, Stop solution 2.