IL-1β (Rat), ELISA Kit
Lieferant: ENZO LIFE SCIENCES
ADI900131A
ENZOADI900131AEA
866
EUR
ENZOADI900131A
IL-1β (Rat), ELISA Kit
Tests
ELISAs
For the quantitative determination of IL-1β in culture supernatants and serum of rat origin.
- Sensitive - use less precious sample per test
- Higher throughput - test up to 40 samples in duplicate in less than 4 h
- Quantitative - obtain statistically significant results and compare data between multiple experiments
- Ready-to-use - pre-coated plates save time and reduce errors
The IL-1β (rat), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in <4 h.
Bestellinformation: Contains microtiter plate, SA-HRP concentrate, SA-HRP dilution buffer, biotinylated antibody, standard, standard diluent, wash buffer concentrate, TMB substrate, stop solution.