IHC tissue primer
Lieferant: ENZO LIFE SCIENCES
ADI9502340100
ENZOADI9502340100EA 234 EUR
ENZOADI9502340100
IHC tissue primer
Tests Immunologische Tests
Immunohistochemistry tissue primer is an enzyme blocking reagent.
  • TRIS-based alkaline solution with 30% H₂O₂
Designed for use in horse radish peroxidase (HRP) and alkaline phosphatase (AP) based immunohistochemical staining procedures on paraffin tissue sections, frozen tissue sections and cell smears to suppress non specific staining due to endogenous enzyme activity.