IHC enzyme antigen retrieval reagent

Lieferant: ENZO LIFE SCIENCES

ADI9502800015
ENZOADI9502800015EA 205 EUR
ENZOADI9502800015
IHC enzyme antigen retrieval reagent
Tests Immunologische Tests

IHC enzyme antigen retrieval reagent is designed to unmask immunoreactive sites altered by fixation and/or the embedding process.


  • Proteolytic enzyme solution that results in superior staining
  • Reagent requires only a brief incubation at room temperature
  • Allow to dilute the primary antibody 5 to 10 fold further and achieve optimal staining


Formulation: Liquid, enzyme antigen retrieval reagent
Appearance: Clear slightly viscous solution.

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR