S-Methyl-L-thiocitrulline dihydrochloride ≥97%
Lieferant: ENZO LIFE SCIENCES
ALX106012M010
ENZOALX106012M010EA
67.1
EUR
ENZOALX106012M010
S-Methyl-L-thiocitrulline dihydrochloride ≥97%
S-Methyl-L-thiocitrulline dihydrochloride
NOS inhibitor
Inhibitor of neuronal nitric oxide synthase (nNOS; NOS I).
CAS-Nummer:
209589-59-3 |