Human TIMP1 (from neutrophils)
Lieferant: ENZO LIFE SCIENCES
ALX200426C005
ENZOALX200426C005EA 299 EUR
ENZOALX200426C005
Human TIMP1 (from neutrophils)
Proteine und Peptide
Isolated from stimulated human neutrophils. The secreted protein consists of 184 amino acids, six disulfide bonds and two glycosylation sites containing N-linked oligosaccharides.
  • Specific inhibitor of matrix metalloproteinases (MMPs)
It has no activity on mammalian membrane metalloproteinases, procollagen peptidase or on bacterial metalloproteinases such as thermolysin.