Phomopsin A ≥98% (durch HPLC)
Lieferant: ENZO LIFE SCIENCES
ALX350417M001
ENZOALX350417M001EA
761
EUR
ENZOALX350417M001
Phomopsin A ≥98% (durch HPLC)
Phomopsin A
Isolated from Phomopsis leptostromiformis MST-FP2068.
A potent mycotoxin and anti-mitotic compound. Causes lupinosis in livestock fed infected lupins. Binds selectively to dimeric tubulin, inhibiting the formation of the microtubule spindle to block cell division. Binds to tubulin at a site overlapping that of vinblastine (Prod. No. ALX-350-257) and maytansine. Protects tubulin from decay.
Formel:
C₃₆H₄₅ClN₆O₁₂ Molecular Weight: 789,2 g/mol |
CAS-Nummer:
64925-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...